5-Nitro-2-(trifluoromethyl)pyrimidine-4,6-diol
Supplier: Matrix Scientific
040949-500MG
101850-484EA
290.77
USD
101850-484
5-Nitro-2-(trifluoromethyl)pyrimidine-4,6-diol
5-Nitro-2-(trifluoromethyl)pyrimidine-4,6-diol
Hazard: Irritant
Formula:
C₅H₂F₃N₃O₄ |
MDL Number:
MFCD08436607 CAS Number: 652-62-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...