6-(Chloromethyl)-2-cyclopropylpyrimidin-4-ol

Supplier: Matrix Scientific

040943-500MG
101850-472EA 387.69 USD
101850-472
6-(Chloromethyl)-2-cyclopropylpyrimidin-4-ol
6-(Chloromethyl)-2-cyclopropylpyrimidin-4-ol

Hazard: Irritant

Formula: C₈H₉ClN₂O
MDL Number: MFCD00084963
CAS Number: 94171-07-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR