6-(Chloromethyl)-2-methylpyrimidin-4-ol

Supplier: Matrix Scientific

040942-500MG
101850-470EA 158.46 USD
101850-470
6-(Chloromethyl)-2-methylpyrimidin-4-ol
6-(Chloromethyl)-2-methylpyrimidin-4-ol

Hazard: Irritant

Formula: C₆H₇ClN₂O
MDL Number: MFCD00129130
CAS Number: 35252-96-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR