2-(2-Aminoethyl)quinazolin-4(3H)-one hydrochloride

Supplier: Matrix Scientific

040939-500MG
101850-464EA 387.69 USD
101850-464
2-(2-Aminoethyl)quinazolin-4(3H)-one hydrochloride
2-(2-Aminoethyl)quinazolin-4(3H)-one hydrochloride

Hazard: Irritant

Formula: C₁₀H₁₂ClN₃O
MDL Number: MFCD11046440
CAS Number: 1185298-40-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR