[1-(Aminomethyl)cyclopentyl]methanol

Supplier: Matrix Scientific

Synonyms: (1-(Aminomethyl)cyclopentyl)methanol

040929-500MG
101850-444EA 290.77 USD
101850-444
[1-(Aminomethyl)cyclopentyl]methanol
[1-(Aminomethyl)cyclopentyl]methanol

Hazard: Irritant

Formula: C₇H₁₅NO
MW: 129.2 g/mol
MDL Number: MFCD09864390
CAS Number: 2239-31-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR