2,4-Dihydroxy-3-methylbenzhydrazide

Supplier: Matrix Scientific

040926-500MG
101850-436EA 217.35 USD
101850-436
2,4-Dihydroxy-3-methylbenzhydrazide
2,4-Dihydroxy-3-methylbenzhydrazide

Hazard: Irritant

Formula: C₈H₁₀N₂O₃
MDL Number: MFCD12028428
CAS Number: 1142211-15-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR