Thiomorpholin-4-ylacetic acid hydrochloride

Supplier: Matrix Scientific

040921-500MG
101850-426EA 387.69 USD
101850-426
Thiomorpholin-4-ylacetic acid hydrochloride
Thiomorpholin-4-ylacetic acid hydrochloride

Hazard: Irritant

Formula: C₆H₁₂ClNO₂S
MDL Number: MFCD12028427
CAS Number: 6007-55-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR