Ethyl thiomorpholin-4-ylacetate

Supplier: Matrix Scientific

040920-500MG
101850-424EA 290.77 USD
101850-424
Ethyl thiomorpholin-4-ylacetate
Ethyl thiomorpholin-4-ylacetate

Hazard: Irritant

Formula: C₈H₁₅NO₂S
MDL Number: MFCD03840689
CAS Number: 39981-80-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR