Methyl-3-(2-ethylpiperidin-1-yl)propanoate

Supplier: Matrix Scientific

040918-500MG
101850-420EA 193.85 USD
101850-420
Methyl-3-(2-ethylpiperidin-1-yl)propanoate
Methyl-3-(2-ethylpiperidin-1-yl)propanoate

Hazard: Irritant

Formula: C₁₁H₂₁NO₂
MDL Number: MFCD11618390
CAS Number: 1094654-27-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR