2-[2-(Methylthio)-1H-benzimidazol-1-yl]propanoic acid

Supplier: Matrix Scientific

040916-500MG
101850-416EA 387.69 USD
101850-416
2-[2-(Methylthio)-1H-benzimidazol-1-yl]propanoic acid
2-[2-(Methylthio)-1H-benzimidazol-1-yl]propanoic acid

Hazard: Irritant

Formula: C₁₁H₁₂N₂O₂S
MDL Number: MFCD09673324
CAS Number: 928712-24-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR