4-(Propionylamino)benzoic acid

Supplier: Matrix Scientific

040902-500MG
101850-398EA 146.15 USD
101850-398
4-(Propionylamino)benzoic acid
4-(Propionylamino)benzoic acid

Hazard: Irritant

Formula: C₁₀H₁₁NO₃
MDL Number: MFCD01365922
CAS Number: 19313-85-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR