5-Isopropylisoxazole-3-carbonyl chloride

Supplier: Matrix Scientific

040897-500MG
101850-388EA 550.77 USD
101850-388
5-Isopropylisoxazole-3-carbonyl chloride
5-Isopropylisoxazole-3-carbonyl chloride

Hazard: Irritant

Formula: C₇H₈ClNO₂
MDL Number: MFCD12028422
CAS Number: 53064-56-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR