5-Ethylisoxazole-3-carbonyl chloride

Supplier: Matrix Scientific

040894-500MG
101850-378EA 360 USD
101850-378
5-Ethylisoxazole-3-carbonyl chloride
5-Ethylisoxazole-3-carbonyl chloride

Hazard: Irritant

Formula: C₆H₆ClNO₂
MDL Number: MFCD12028420
CAS Number: 52320-58-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR