4-Chloro-1-ethyl-1H-pyrazole-5-carbonyl chloride

Supplier: Matrix Scientific

040893-500MG
101850-376EA 404.62 USD
101850-376
4-Chloro-1-ethyl-1H-pyrazole-5-carbonyl chloride
4-Chloro-1-ethyl-1H-pyrazole-5-carbonyl chloride

Hazard: Irritant

Formula: C₆H₆Cl₂N₂O
MDL Number: MFCD03421438
CAS Number: 1171932-05-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR