4-Bromo-1-methyl-1H-pyrazole-5-carbonyl chloride

Supplier: Matrix Scientific

040892-500MG
101850-374EA 484.62 USD
101850-374
4-Bromo-1-methyl-1H-pyrazole-5-carbonyl chloride
4-Bromo-1-methyl-1H-pyrazole-5-carbonyl chloride

Hazard: Irritant

Formula: C₅H₄BrClN₂O
MDL Number: MFCD03421425
CAS Number: 1006448-72-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR