Ethyl-5-(2-furyl)-7-(trifluoromethyl)-4,5,6,7-tetrahydropyrazolo[1,5-a]pyrimidine-3-carboxylate

Supplier: Matrix Scientific

040887-500MG
101850-364EA 387.69 USD
101850-364
Ethyl-5-(2-furyl)-7-(trifluoromethyl)-4,5,6,7-tetrahydropyrazolo[1,5-a]pyrimidine-3-carboxylate
Ethyl-5-(2-furyl)-7-(trifluoromethyl)-4,5,6,7-tetrahydropyrazolo[1,5-a]pyrimidine-3-carboxylate

Hazard: Irritant

Formula: C₁₄H₁₄F₃N₃O₃
MDL Number: MFCD04967320
CAS Number: 667920-77-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR