Methyl-5-(2-furyl)-7-(trifluoromethyl)pyrazolo[1,5-a]pyrimidine-2-carboxylate
Supplier: Matrix Scientific
040883-500MG
101850-358EA
290.77
USD
101850-358
Methyl-5-(2-furyl)-7-(trifluoromethyl)pyrazolo[1,5-a]pyrimidine-2-carboxylate
Methyl-5-(2-furyl)-7-(trifluoromethyl)pyrazolo[1,5-a]pyrimidine-2-carboxylate
Hazard: Irritant
Formula:
C₁₃H₈F₃N₃O₃ |
MDL Number:
MFCD12028419 CAS Number: 1142211-08-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...