Ethyl-5-(2-thienyl)-7-(trifluoromethyl)-4,5,6,7-tetrahydropyrazolo[1,5-a]pyrimidine-3-carboxylate

Supplier: Matrix Scientific

040880-500MG
101850-352EA 387.69 USD
101850-352
Ethyl-5-(2-thienyl)-7-(trifluoromethyl)-4,5,6,7-tetrahydropyrazolo[1,5-a]pyrimidine-3-carboxylate
Ethyl-5-(2-thienyl)-7-(trifluoromethyl)-4,5,6,7-tetrahydropyrazolo[1,5-a]pyrimidine-3-carboxylate

Hazard: Irritant

Formula: C₁₄H₁₄F₃N₃O₂S
MDL Number: MFCD04967332
CAS Number: 827592-52-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR