Ethyl (4-bromo-5-methyl-3-nitro-1H-pyrazol-1-yl)acetate

Supplier: Matrix Scientific

Synonyms: Ethyl 2-(4-bromo-5-methyl-3-nitro-1H-pyrazol-1-yl)acetate

040877-500MG
101850-346EA 387.69 USD
101850-346
Ethyl (4-bromo-5-methyl-3-nitro-1H-pyrazol-1-yl)acetate
Ethyl (4-bromo-5-methyl-3-nitro-1H-pyrazol-1-yl)acetate

Hazard: Irritant

Formula: C₈H₁₀BrN₃O₄
MW: 292.1 g/mol
MDL Number: MFCD00682561
CAS Number: 351992-36-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR