7-(Difluoromethyl)-5-(4-fluorophenyl)pyrazolo[1,5-a]pyrimidine-3-carboxylic acid

Supplier: Matrix Scientific

040874-500MG
101850-340EA 290.77 USD
101850-340
7-(Difluoromethyl)-5-(4-fluorophenyl)pyrazolo[1,5-a]pyrimidine-3-carboxylic acid
7-(Difluoromethyl)-5-(4-fluorophenyl)pyrazolo[1,5-a]pyrimidine-3-carboxylic acid

Hazard: Irritant

Formula: C₁₄H₈F₃N₃O₂
MW: 307.23 g/mol
MDL Number: MFCD04967191
CAS Number: 712319-09-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR