5-(4-Fluorophenyl)-2-methyl-2H-1,2,6-thiadiazine-3-carboxylic acid 1,1-dioxide

Supplier: Matrix Scientific

040867-500MG
101850-328EA 290.77 USD
101850-328
5-(4-Fluorophenyl)-2-methyl-2H-1,2,6-thiadiazine-3-carboxylic acid 1,1-dioxide
5-(4-Fluorophenyl)-2-methyl-2H-1,2,6-thiadiazine-3-carboxylic acid 1,1-dioxide

Hazard: Irritant

Formula: C₁₁H₉FN₂O₄S
MDL Number: MFCD08106851
CAS Number: 899539-96-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR