5-(2,5-Dimethoxyphenyl)-2-methyl-2H-1,2,6-thiadiazine-3-carboxylic acid-1,1-dioxide
Supplier: Matrix Scientific
040866-500MG
101850-326EA
290.77
USD
101850-326
5-(2,5-Dimethoxyphenyl)-2-methyl-2H-1,2,6-thiadiazine-3-carboxylic acid-1,1-dioxide
5-(2,5-Dimethoxyphenyl)-2-methyl-2H-1,2,6-thiadiazine-3-carboxylic acid-1,1-dioxide
Hazard: Irritant
Formula:
C₁₃H₁₄N₂O₆S MW: 326.33 g/mol |
MDL Number:
MFCD08106872 CAS Number: 899540-36-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...