1-Ethyl-5-methyl-4-nitro-1H-pyrazol-3-amine

Supplier: Matrix Scientific

040855-500MG
101850-306EA 484.62 USD
101850-306
1-Ethyl-5-methyl-4-nitro-1H-pyrazol-3-amine
1-Ethyl-5-methyl-4-nitro-1H-pyrazol-3-amine

Hazard: Irritant

Formula: C₆H₁₀N₄O₂
MDL Number: MFCD12028413
CAS Number: 1170024-12-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR