3-(2,5-Dimethoxyphenyl)-4,5-dihydroisoxazole-5-carboxylic acid

Supplier: Matrix Scientific

040853-500MG
101850-302EA 484.62 USD
101850-302
3-(2,5-Dimethoxyphenyl)-4,5-dihydroisoxazole-5-carboxylic acid
3-(2,5-Dimethoxyphenyl)-4,5-dihydroisoxazole-5-carboxylic acid

Hazard: Irritant

Formula: C₁₂H₁₃NO₅
MDL Number: MFCD09701761
CAS Number: 1018143-27-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR