3-Chloro-6-methylpyrazolo[1,5-a]pyrimidine-2-carboxylic acid

Supplier: Matrix Scientific

040850-500MG
101850-296EA 146.15 USD
101850-296
3-Chloro-6-methylpyrazolo[1,5-a]pyrimidine-2-carboxylic acid
3-Chloro-6-methylpyrazolo[1,5-a]pyrimidine-2-carboxylic acid

Hazard: Irritant

Formula: C₈H₆ClN₃O₂
MDL Number: MFCD09971313
CAS Number: 1015846-76-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR