5-(4-Fluorophenyl)-7-(trifluoromethyl)pyrazolo[1,5-a]pyrimidine-2-carboxylic acid

Supplier: Matrix Scientific

040845-500MG
101850-286EA 290.77 USD
101850-286
5-(4-Fluorophenyl)-7-(trifluoromethyl)pyrazolo[1,5-a]pyrimidine-2-carboxylic acid
5-(4-Fluorophenyl)-7-(trifluoromethyl)pyrazolo[1,5-a]pyrimidine-2-carboxylic acid

Hazard: Irritant

Formula: C₁₄H₇F₄N₃O₂
MW: 325.22 g/mol
MDL Number: MFCD01194316
CAS Number: 310451-82-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR