7-(Difluoromethyl)-5-methyl-4,5,6,7-tetrahydropyrazolo[1,5-a]pyrimidine-2-carboxylic acid
Supplier: Matrix Scientific
040841-500MG
101850-282EA
484.62
USD
101850-282
7-(Difluoromethyl)-5-methyl-4,5,6,7-tetrahydropyrazolo[1,5-a]pyrimidine-2-carboxylic acid
7-(Difluoromethyl)-5-methyl-4,5,6,7-tetrahydropyrazolo[1,5-a]pyrimidine-2-carboxylic acid
Hazard: Irritant
Formula:
C₉H₁₁F₂N₃O₂ MW: 231.2 g/mol |
MDL Number:
MFCD04967190 CAS Number: 725699-03-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...