7-(Difluoromethyl)-5-phenylpyrazolo[1,5-a]pyrimidine-3-carboxylic acid

Supplier: Matrix Scientific

040838-500MG
101850-276EA 387.69 USD
101850-276
7-(Difluoromethyl)-5-phenylpyrazolo[1,5-a]pyrimidine-3-carboxylic acid
7-(Difluoromethyl)-5-phenylpyrazolo[1,5-a]pyrimidine-3-carboxylic acid

Hazard: Irritant

Formula: C₁₄H₉F₂N₃O₂
MW: 289.24 g/mol
MDL Number: MFCD02253717
CAS Number: 438220-85-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR