7-[Chloro(difluoro)methyl]-5-methylpyrazolo[1,5-a]pyrimidine-2-carboxylic acid

Supplier: Matrix Scientific

040831-500MG
101850-262EA 387.69 USD
101850-262
7-[Chloro(difluoro)methyl]-5-methylpyrazolo[1,5-a]pyrimidine-2-carboxylic acid
7-[Chloro(difluoro)methyl]-5-methylpyrazolo[1,5-a]pyrimidine-2-carboxylic acid

Hazard: Irritant

Formula: C₉H₆ClF₂N₃O₂
MW: 261.62 g/mol
MDL Number: MFCD04045223
CAS Number: 676522-75-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR