Ethyl-7-(difluoromethyl)-5-(2-methoxyphenyl)pyrazolo[1,5-a]pyrimidine-3-carboxylate

Supplier: Matrix Scientific

040830-500MG
101850-260EA 290.77 USD
101850-260
Ethyl-7-(difluoromethyl)-5-(2-methoxyphenyl)pyrazolo[1,5-a]pyrimidine-3-carboxylate
Ethyl-7-(difluoromethyl)-5-(2-methoxyphenyl)pyrazolo[1,5-a]pyrimidine-3-carboxylate

Hazard: Irritant

Formula: C₁₇H₁₅F₂N₃O₃
MDL Number: MFCD04048850
CAS Number: 754230-33-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR