Ethyl-7-(difluoromethyl)-5-(4-methylphenyl)pyrazolo[1,5-a]pyrimidine-3-carboxylate

Supplier: Matrix Scientific

040829-500MG
101850-258EA 290.77 USD
101850-258
Ethyl-7-(difluoromethyl)-5-(4-methylphenyl)pyrazolo[1,5-a]pyrimidine-3-carboxylate
Ethyl-7-(difluoromethyl)-5-(4-methylphenyl)pyrazolo[1,5-a]pyrimidine-3-carboxylate

Hazard: Irritant

Formula: C₁₇H₁₅F₂N₃O₂
MDL Number: MFCD03074470
CAS Number: 438218-13-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR