Methyl-4-bromo-1-methyl-1H-pyrazole-5-carboxylate

Supplier: Matrix Scientific

Synonyms: Methyl 4-bromo-1-methyl-1H-pyrazole-5-carboxylate

040828-500MG
101850-256EA 95.38 USD
101850-256
Methyl-4-bromo-1-methyl-1H-pyrazole-5-carboxylate
Methyl-4-bromo-1-methyl-1H-pyrazole-5-carboxylate

Hazard: Irritant

Formula: C₆H₇BrN₂O₂
MW: 219.04 g/mol
MDL Number: MFCD01459850
CAS Number: 514816-42-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR