[(3-Ethyl-4,5-dihydroisoxazol-5-yl)methyl]amine hydrochloride

Supplier: Matrix Scientific

040827-500MG
101850-254EA 375.38 USD
101850-254
[(3-Ethyl-4,5-dihydroisoxazol-5-yl)methyl]amine hydrochloride
[(3-Ethyl-4,5-dihydroisoxazol-5-yl)methyl]amine hydrochloride

Hazard: Irritant

Formula: C₆H₁₃ClN₂O
MDL Number: MFCD12028412
CAS Number: 1160245-60-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR