[3-(2-Methylpyrazolo[1,5-a]pyrimidin-7-yl)benzyl]amine hydrochloride

Supplier: Matrix Scientific

040825-500MG
101850-250EA 335.38 USD
101850-250
[3-(2-Methylpyrazolo[1,5-a]pyrimidin-7-yl)benzyl]amine hydrochloride
[3-(2-Methylpyrazolo[1,5-a]pyrimidin-7-yl)benzyl]amine hydrochloride

Hazard: Irritant

Formula: C₁₄H₁₅ClN₄
MDL Number: MFCD12028410
CAS Number: 1185294-93-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR