1,5-Dimethyl-1H-pyrazol-4-amine hydrochloride

Supplier: Matrix Scientific

Synonyms: 1,5-Dimethyl-1H-pyrazol-4-amine xhydrochloride

040821-500MG
101850-242EA 484.62 USD
101850-242
1,5-Dimethyl-1H-pyrazol-4-amine hydrochloride
1,5-Dimethyl-1H-pyrazol-4-amine hydrochloride

Hazard: Irritant

Formula: C₅H₁₀ClN₃
MW: 147.61 g/mol
MDL Number: MFCD11100176
CAS Number: 948571-57-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR