5-(3-Methoxyphenyl)-7-(trifluoromethyl)pyrazolo[1,5-a]pyrimidine-2-carboxylic acid

Supplier: Matrix Scientific

040818-500MG
101850-236EA 129.23 USD
101850-236
5-(3-Methoxyphenyl)-7-(trifluoromethyl)pyrazolo[1,5-a]pyrimidine-2-carboxylic acid
5-(3-Methoxyphenyl)-7-(trifluoromethyl)pyrazolo[1,5-a]pyrimidine-2-carboxylic acid

Hazard: Irritant

Formula: C₁₅H₁₀F₃N₃O₃
MW: 337.26 g/mol
MDL Number: MFCD02253965
CAS Number: 524036-15-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR