3-Chloro-5-cyclopropyl-7-(trifluoromethyl)pyrazolo[1,5-a]pyrimidine-2-carboxylic acid

Supplier: Matrix Scientific

040814-500MG
101850-228EA 335.38 USD
101850-228
3-Chloro-5-cyclopropyl-7-(trifluoromethyl)pyrazolo[1,5-a]pyrimidine-2-carboxylic acid
3-Chloro-5-cyclopropyl-7-(trifluoromethyl)pyrazolo[1,5-a]pyrimidine-2-carboxylic acid

Hazard: Irritant

Formula: C₁₁H₇ClF₃N₃O₂
MDL Number: MFCD05239177
CAS Number: 797809-11-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR