3-Chloro-5-methyl-7-(trifluoromethyl)pyrazolo[1,5-a]pyrimidine-2-carboxylic acid

Supplier: Matrix Scientific

040811-500MG
101850-222EA 387.69 USD
101850-222
3-Chloro-5-methyl-7-(trifluoromethyl)pyrazolo[1,5-a]pyrimidine-2-carboxylic acid
3-Chloro-5-methyl-7-(trifluoromethyl)pyrazolo[1,5-a]pyrimidine-2-carboxylic acid

Hazard: Irritant

Formula: C₉H₅ClF₃N₃O₂
MDL Number: MFCD02253966
CAS Number: 524036-11-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR