3-Chloro-5-phenyl-7-(trifluoromethyl)-4,5,6,7-tetrahydropyrazolo[1,5-a]pyrimidine-2-carboxylic acid

Supplier: Matrix Scientific

040807-500MG
101850-214EA 192.31 USD
101850-214
3-Chloro-5-phenyl-7-(trifluoromethyl)-4,5,6,7-tetrahydropyrazolo[1,5-a]pyrimidine-2-carboxylic acid
3-Chloro-5-phenyl-7-(trifluoromethyl)-4,5,6,7-tetrahydropyrazolo[1,5-a]pyrimidine-2-carboxylic acid

Hazard: Irritant

Formula: C₁₄H₁₁ClF₃N₃O₂
MDL Number: MFCD01922903
CAS Number: 312699-22-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR