3-Chloro-5-phenyl-7-(trifluoromethyl)pyrazolo[1,5-a]pyrimidine-2-carboxylic acid

Supplier: Matrix Scientific

040797-500MG
101850-198EA 290.77 USD
101850-198
3-Chloro-5-phenyl-7-(trifluoromethyl)pyrazolo[1,5-a]pyrimidine-2-carboxylic acid
3-Chloro-5-phenyl-7-(trifluoromethyl)pyrazolo[1,5-a]pyrimidine-2-carboxylic acid

Hazard: Irritant

Formula: C₁₄H₇ClF₃N₃O₂
MDL Number: MFCD00301491
CAS Number: 299405-80-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR