5-(2-Methoxyphenyl)-7-(trifluoromethyl)pyrazolo[1,5-a]pyrimidine-3-carboxylic acid

Supplier: Matrix Scientific

040796-500MG
101850-196EA 290.77 USD
101850-196
5-(2-Methoxyphenyl)-7-(trifluoromethyl)pyrazolo[1,5-a]pyrimidine-3-carboxylic acid
5-(2-Methoxyphenyl)-7-(trifluoromethyl)pyrazolo[1,5-a]pyrimidine-3-carboxylic acid

Hazard: Irritant

Formula: C₁₅H₁₀F₃N₃O₃
MW: 337.26 g/mol
MDL Number: MFCD06626674
CAS Number: 831229-13-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR