6-Bromo-3-chloropyrazolo[1,5-a]pyrimidine-2-carboxylic acid

Supplier: Matrix Scientific

040793-500MG
101850-190EA 170.77 USD
101850-190
6-Bromo-3-chloropyrazolo[1,5-a]pyrimidine-2-carboxylic acid
6-Bromo-3-chloropyrazolo[1,5-a]pyrimidine-2-carboxylic acid

Hazard: Irritant

Formula: C₇H₃BrClN₃O₂
MDL Number: MFCD00297265
CAS Number: 499190-16-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR