5-(3-Methoxyphenyl)-7-(trifluoromethyl)pyrazolo[1,5-a]pyrimidine-3-carboxylic acid

Supplier: Matrix Scientific

040792-500MG
101850-188EA 290.77 USD
101850-188
5-(3-Methoxyphenyl)-7-(trifluoromethyl)pyrazolo[1,5-a]pyrimidine-3-carboxylic acid
5-(3-Methoxyphenyl)-7-(trifluoromethyl)pyrazolo[1,5-a]pyrimidine-3-carboxylic acid

Hazard: Irritant

Formula: C₁₅H₁₀F₃N₃O₃
MW: 337.26 g/mol
MDL Number: MFCD02253979
CAS Number: 797809-13-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR