3,5-Dimethyl-4-pyrrolidin-2-ylisoxazole

Supplier: Matrix Scientific

040782-500MG
101850-168EA 484.62 USD
101850-168
3,5-Dimethyl-4-pyrrolidin-2-ylisoxazole
3,5-Dimethyl-4-pyrrolidin-2-ylisoxazole

Hazard: Irritant

Formula: C₉H₁₄N₂O
MDL Number: MFCD09859204
CAS Number: 1018128-26-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR