2-(2-Chlorophenyl)pyrrolidine

Supplier: Matrix Scientific

040777-500MG
101850-158EA 484.62 USD
101850-158
2-(2-Chlorophenyl)pyrrolidine
2-(2-Chlorophenyl)pyrrolidine

Hazard: Irritant

Formula: C₁₀H₁₂ClN
MDL Number: MFCD02089473
CAS Number: 129540-21-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR