[3-(4-Chlorophenyl)-5-isoxazolyl]methanamine

Supplier: Matrix Scientific

040775-500MG
101850-154EA 300 USD
101850-154
[3-(4-Chlorophenyl)-5-isoxazolyl]methanamine
[3-(4-Chlorophenyl)-5-isoxazolyl]methanamine

Hazard: Irritant

Formula: C₁₀H₉ClN₂O
MDL Number: MFCD01935956
CAS Number: 66046-42-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR