Ethyl-4-methyl-1,2,3-thiadiazole-5-carboxylate

Supplier: Matrix Scientific

Synonyms: Ethyl 4-methyl-1,2,3-thiadiazole-5-carboxylate

040773-500MG
101850-150EA 387.69 USD
101850-150
Ethyl-4-methyl-1,2,3-thiadiazole-5-carboxylate
Ethyl-4-methyl-1,2,3-thiadiazole-5-carboxylate

Hazard: Irritant

Formula: C₆H₈N₂O₂S
MW: 172.21 g/mol
MDL Number: MFCD00052549
CAS Number: 18212-20-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR