Methyl-3-ethyl-4,5-dihydroisoxazole-5-carboxylate

Supplier: Matrix Scientific

040770-500MG
101850-144EA 387.69 USD
101850-144
Methyl-3-ethyl-4,5-dihydroisoxazole-5-carboxylate
Methyl-3-ethyl-4,5-dihydroisoxazole-5-carboxylate

Hazard: Irritant

Formula: C₇H₁₁NO₃
MDL Number: MFCD12028404
CAS Number: 55134-84-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR