Ethyl-3-ethyl-5-methylisoxazole-4-carboxylate

Supplier: Matrix Scientific

040766-500MG
101850-136EA 387.69 USD
101850-136
Ethyl-3-ethyl-5-methylisoxazole-4-carboxylate
Ethyl-3-ethyl-5-methylisoxazole-4-carboxylate

Hazard: Irritant

Formula: C₉H₁₃NO₃
MDL Number: MFCD00014094
CAS Number: 53064-41-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR