2-(3-Methyl-2-thienyl)azepane

Supplier: Matrix Scientific

040761-500MG
101850-124EA 484.62 USD
101850-124
2-(3-Methyl-2-thienyl)azepane
2-(3-Methyl-2-thienyl)azepane

Hazard: Irritant

Formula: C₁₁H₁₇NS
MDL Number: MFCD05190318
CAS Number: 527674-19-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR